Gene Transcripts

Gene Symbol
Sprr2a3
Entrez Gene ID
100042514
Gene Name
small proline-rich protein 2A3
Gene Aliases
Chromosome Location
Chr. 3: 92284658 - 92289255 on Build GRCm38
UniGene ID
Mm.359329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC051122.1 - 1 202
BC056231.1 - 1 213
BC099393.1 - 1 231 30790331

Gene Symbol
Sprr2a1
Entrez Gene ID
20755
Gene Name
small proline-rich protein 2A1
Gene Aliases
Sprr2a, Sprr2a3
Chromosome Location
Chr. 3: 92215835 - 92222491 on Build GRCm38
UniGene ID
Mm.6853
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011468.4 NP_035598.2 2 327
GenBank mRNA AK003391.1 - 2 243
AK007633.1 - 2 243
AK008631.1 - 2 240
AK008887.1 - 2 243
AK009790.1 - 2 240
AK147183.1 - 2 243
AK168529.1 - 2 242
AY158986.1 - 240

Gene Symbol
Sprr2a2
Entrez Gene ID
100303744
Gene Name
small proline-rich protein 2A2
Gene Aliases
Chromosome Location
Chr. 3: 92250647 - 92257304 on Build GRCm38
UniGene ID
Mm.429851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164787.1 NP_001158259.1 327
GenBank mRNA AK007928.1 - 238
BC010818.1 - 227 4216389

View Full Product Details