Gene Transcripts

Gene Symbol
MRPS10
Entrez Gene ID
55173
Gene Name
mitochondrial ribosomal protein S10
Gene Aliases
MRP-S10, PNAS-122
Chromosome Location
Chr. 6: 42206801 - 42217907 on Build GRCh38
UniGene ID
Hs.380887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018141.3 NP_060611.2 2 135
XM_005249206.2 XP_005249263.1 2 147
XM_011514724.2 XP_011513026.1 3 139
XM_011514725.2 XP_011513027.1 3 134
XM_017010996.1 XP_016866485.1 2 110
GenBank mRNA AF113220.1 - 2 95
AF277180.1 - 2 97
AK001429.1 - 2 104
AK098446.1 - 2 98
AK298219.1 - 2 92
AK316599.1 - 2 101
BC012560.1 - 2 105 4277803
BG776014.1 - 2 136

View Full Product Details