Gene Transcripts

Gene Symbol
GGACT
Entrez Gene ID
87769
Gene Name
gamma-glutamylamine cyclotransferase
Gene Aliases
A2LD1
Chromosome Location
Chr. 13: 100530164 - 100588792 on Build GRCh38
UniGene ID
Hs.350868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033110.2 NP_149101.1 1 50
XM_011521129.2 XP_011519431.1 2 3013
GenBank mRNA DA628207.1 - 1 50

View Full Product Details