Gene Transcripts

Gene Symbol
TSPAN19
Entrez Gene ID
144448
Gene Name
tetraspanin 19
Gene Aliases
Chromosome Location
Chr. 12: 85014311 - 85036277 on Build GRCh38
UniGene ID
Hs.156962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100917.1 NP_001094387.1 2, 3 167
XM_005268669.4 XP_005268726.1 2, 3 217
XM_017018862.1 XP_016874351.1 1, 2 105
XM_017018863.1 XP_016874352.1 1, 2 105
XM_017018864.1 XP_016874353.1 1, 2 105
GenBank mRNA AK126325.1 - 2, 3 167
BX116902.1 - 2, 3 147

View Full Product Details