Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
Chromosome Location
Chr. 19: 4182618 - 4224814 on Build GRCh38
UniGene ID
Hs.197872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133475.1 NP_597732.1 20 3551
XM_011527756.2 XP_011526058.1 22 3656
XM_011527757.2 XP_011526059.1 20 3709
XM_011527758.2 XP_011526060.1 21 3569
XM_011527761.1 XP_011526063.1 23 3726
XM_011527764.2 XP_011526066.1 19 3622
XM_011527765.2 XP_011526067.1 20 3470
XM_011527768.2 XP_011526070.1 8 2427
XM_017026438.1 XP_016881927.1 23 3717
XM_017026439.1 XP_016881928.1 22 3639
XM_017026440.1 XP_016881929.1 21 3612
XM_017026441.1 XP_016881930.1 11 2754

View Full Product Details