Gene Transcripts

Gene Symbol
PATL2
Entrez Gene ID
197135
Gene Name
PAT1 homolog 2
Gene Aliases
Pat1a, hPat1a
Chromosome Location
Chr. 15: 44665732 - 44711389 on Build GRCh38
UniGene ID
Hs.11594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145112.1 NP_001138584.1 10 1008
XM_011521336.2 XP_011519638.2 10 1834
XM_011521337.2 XP_011519639.2 10 1969
XM_011521338.2 XP_011519640.1 12 1533
XM_011521339.2 XP_011519641.1 11 1414
XM_011521340.2 XP_011519642.1 10 1355
XM_011521341.1 XP_011519643.1 10 1118
XM_011521342.2 XP_011519644.1 8 2015
XM_011521343.2 XP_011519645.1 11 1365
XM_011521344.2 XP_011519646.1 8 807
XM_011521345.2 XP_011519647.1 11 1327
XM_011521346.2 XP_011519648.2 6 1541
XM_011521347.1 XP_011519649.1 9 923
XM_011521348.2 XP_011519650.1 9 923
XM_017022000.1 XP_016877489.1 10 1826
XM_017022001.1 XP_016877490.1 8 955
GenBank mRNA BI909158.1 - 2 108

View Full Product Details