Gene Transcripts
- Gene Symbol
- PABPC1L
- Entrez Gene ID
- 80336
- Gene Name
- poly(A) binding protein cytoplasmic 1 like
- Gene Aliases
- C20orf119, EPAB, PABPC1L1, dJ1069P2.3
- Chromosome Location
- Chr. 20: 44910060 - 44959160 on Build GRCh38
- UniGene ID
- Hs.641481
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001124756.2 |
NP_001118228.1
|
12, 13 |
1721 |
|
NR_134983.1 |
|
11, 12 |
1572 |
|
NR_134984.1 |
|
3, 4 |
1817 |
|
NR_134985.1 |
|
3, 4 |
1817 |
|
NR_134986.1 |
|
3, 4 |
1817 |
|
GenBank mRNA |
AK026760.1 |
- |
3, 4 |
1200 |
|
AK124047.1 |
- |
3, 4 |
1817 |
|
BC068104.1 |
- |
3, 4 |
1001 |
|
BM832739.1 |
- |
3, 4 |
278 |
|
EU190483.1 |
- |
11, 12 |
1508 |
|
View Full Product Details