Gene Transcripts

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
Chromosome Location
Chr. 5: 132750817 - 132778216 on Build GRCh38
UniGene ID
Hs.522057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098811.1 NP_001092281.1 10 2704
XM_005271927.2 XP_005271984.1 10 2703
XM_005271928.2 XP_005271985.1 10 2700
XM_005271931.3 XP_005271988.1 11 2787
XM_011543288.1 XP_011541590.1 10 2726
XM_017009254.1 XP_016864743.1 10 2697
XM_017009255.1 XP_016864744.1 10 2770
XM_017009256.1 XP_016864745.1 10 2767
XM_017009257.1 XP_016864746.1 10 2720
GenBank mRNA AF440762.1 - 10 2704

View Full Product Details