Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146339.1 | NP_001139811.1 | 5 | 1163 | |
XM_005258856.3 | XP_005258913.1 | 3 | 1086 | ||
XM_011526902.2 | XP_011525204.1 | 6 | 1783 | ||
XM_011526903.2 | XP_011525205.1 | 5 | 1753 | ||
XM_011526904.2 | XP_011525206.1 | 5 | 1308 | ||
XM_011526905.2 | XP_011525207.1 | 4 | 1158 |