Gene Transcripts

Gene Symbol
OGDHL
Entrez Gene ID
55753
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
Chromosome Location
Chr. 10: 49734641 - 49762379 on Build GRCh38
UniGene ID
Hs.17860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143996.1 NP_001137468.1 4 474
NM_001143997.1 NP_001137469.1 3 269
NM_018245.2 NP_060715.2 5 645
XM_011539946.1 XP_011538248.1 5 530
XM_011539947.1 XP_011538249.1 5 736
XM_017016401.1 XP_016871890.1 5 920
XM_017016402.1 XP_016871891.1 5 520
XM_017016403.1 XP_016871892.1 4 384
GenBank mRNA AB033116.3 - 4 508
AK001713.1 - 5 589
AK225558.1 - 5 576
AK290226.1 - 4 618
AK296551.1 - 4 383
AK303729.1 - 3 216
AK307759.1 - 4 404
BC026320.1 - 5 564 4797814
DA411851.1 - 1 37
DA416844.1 - 2 55

View Full Product Details