Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
Chromosome Location
Chr. 19: 45995457 - 46018616 on Build GRCh38
UniGene ID
Hs.515479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267723.1 NP_001254652.1 11 1337
XM_005259191.3 XP_005259248.1 11 3442
XM_005259192.4 XP_005259249.1 11 2083
XM_011527257.2 XP_011525559.1 11 1366
GenBank mRNA CA775840.1 - 3 275
CB852675.1 - 4 324
GQ375050.1 - 7 770

View Full Product Details