Gene Transcripts
- Gene Symbol
- TMEM39A
- Entrez Gene ID
- 55254
- Gene Name
- transmembrane protein 39A
- Gene Aliases
- Chromosome Location
- Chr. 3: 119428960 - 119463682 on Build GRCh38
- UniGene ID
- Hs.594171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018266.2 |
NP_060736.1
|
2, 3 |
542 |
|
NR_073506.1 |
|
2, 3 |
542 |
|
XM_005247578.1 |
XP_005247635.1
|
2, 3 |
507 |
|
XM_006713687.1 |
XP_006713750.1
|
2, 3 |
450 |
|
XM_011512958.2 |
XP_011511260.1
|
2, 3 |
507 |
|
XM_017006788.1 |
XP_016862277.1
|
2, 3 |
507 |
|
GenBank mRNA |
AK001764.1 |
- |
2, 3 |
484 |
|
AK222950.1 |
- |
2, 3 |
474 |
|
AK223248.1 |
- |
2, 3 |
474 |
|
BC003192.1 |
- |
2, 3 |
457 |
|
BC010070.2 |
- |
2, 3 |
457 |
|
BC021277.2 |
- |
2, 3 |
460 |
4857987 |
View Full Product Details