Gene Transcripts

Gene Symbol
UEVLD
Entrez Gene ID
55293
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
ATTP, UEV3
Chromosome Location
Chr. 11: 18529609 - 18588837 on Build GRCh38
UniGene ID
Hs.407991
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040697.2 NP_001035787.1 12 1645
NM_001261382.1 NP_001248311.1 11 1579
NM_001261383.1 NP_001248312.1 10 1455
NM_001261384.1 NP_001248313.1 10 1396
NM_001261385.1 NP_001248314.1 11 1478
NM_001261386.1 NP_001248315.1 7 986
NM_018314.4 NP_060784.3 11 1521
XM_006718255.2 XP_006718318.1 11 1474
XM_011520201.2 XP_011518503.1 8 3476
XM_017017985.1 XP_016873474.1 11 1572
XM_017017986.1 XP_016873475.1 10 1328
GenBank mRNA AF503350.1 - 11 1481
AK001930.1 - 11 1478
AK295501.1 - 10 1324
AK296835.1 - 11 1539
AL134932.1 - 1 286
BC064566.1 - 10 1395 5576632

View Full Product Details