Gene Transcripts

Gene Symbol
C2orf72
Entrez Gene ID
257407
Gene Name
chromosome 2 open reading frame 72
Gene Aliases
Chromosome Location
Chr. 2: 231037566 - 231049714 on Build GRCh38
UniGene ID
Hs.526596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144994.1 NP_001138466.1 3 1125
GenBank mRNA AF007149.1 - 2 612

View Full Product Details