Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018445.5 | NP_060915.2 | 4 | 446 | |
NM_203472.2 | NP_982298.2 | 4 | 446 | ||
GenBank mRNA | AF157317.1 | - | 4 | 373 | |
AF328864.1 | - | 4 | 373 | ||
AK026455.1 | - | 4 | 409 | ||
AK097011.1 | - | 2 | 1636 | ||
AK225955.1 | - | 4 | 372 | ||
AK314723.1 | - | 4 | 408 | ||
AY324824.1 | - | 4 | 373 | ||
AY618665.1 | - | 4 | 343 | ||
BC005840.2 | - | 4 | 369 | 2967406 | |
BC107774.1 | - | 4 | 376 | 6450503 | |
BG829960.1 | - | 4 | 446 |