Gene Transcripts

Gene Symbol
SHISA9
Entrez Gene ID
729993
Gene Name
shisa family member 9
Gene Aliases
CKAMP44
Chromosome Location
Chr. 16: 12901619 - 13563427 on Build GRCh38
UniGene ID
Hs.130661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145205.1 NP_001138677.2 2 1044
GenBank mRNA AJ002784.1 - 1 63
AK309038.1 - 2 385
CR627300.1 - 2 240
CR749545.1 - 2 240

View Full Product Details