Gene Transcripts
- Gene Symbol
- SLC37A3
- Entrez Gene ID
- 84255
- Gene Name
- solute carrier family 37 member 3
- Gene Aliases
- Chromosome Location
- Chr. 7: 140333752 - 140404352 on Build GRCh38
- UniGene ID
- Hs.446021
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131347.1 |
- |
2 |
1563 |
|
View Full Product Details