Gene Transcripts

Gene Symbol
PRSS51
Entrez Gene ID
346702
Gene Name
protease, serine 51
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.675958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK307207.1 - 5 851

View Full Product Details