Gene Transcripts

Gene Symbol
TRABD2A
Entrez Gene ID
129293
Gene Name
TraB domain containing 2A
Gene Aliases
C2orf89, TIKI1
Chromosome Location
Chr. 2: 84821650 - 84881975 on Build GRCh38
UniGene ID
Hs.469134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080824.2 NP_001074293.1 2 331
NM_001277053.1 NP_001263982.1 2 331
NM_001307978.1 NP_001294907.1 2 331
XM_011532504.1 XP_011530806.1 2 131
GenBank mRNA AK296608.1 - 2 168
AK309615.1 - 2 170
BC051789.1 - 2 332
JQ653415.1 - 2 125

View Full Product Details