Gene Transcripts

Gene Symbol
PWRN2
Entrez Gene ID
791115
Gene Name
Prader-Willi region non-protein coding RNA 2
Gene Aliases
NCRNA00199
Chromosome Location
Chr. 15: 24164779 - 24169906 on Build GRCh38
UniGene ID
Hs.560378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026647.1 1 2085
GenBank mRNA AK124131.1 - 2 1250
BC032951.1 - 2 1211
BC070120.1 - 2 1206

View Full Product Details