Gene Transcripts

Gene Symbol
RHOF
Entrez Gene ID
54509
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
ARHF, RIF
Chromosome Location
Chr. 12: 121777754 - 121793688 on Build GRCh38
UniGene ID
Hs.707579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019034.2 NP_061907.2 5 2317
GenBank mRNA AK000254.1 - 5 2285

View Full Product Details