Gene Transcripts
- Gene Symbol
- KRTAP2-2
- Entrez Gene ID
- 728279
- Gene Name
- keratin associated protein 2-2
- Gene Aliases
- KAP2.2
- Chromosome Location
- Chr. 17: 41054498 - 41055211 on Build GRCh38
- UniGene ID
- Hs.650131
- Species
- Homo sapiens
- Gene Symbol
- KRTAP2-4
- Entrez Gene ID
- 85294
- Gene Name
- keratin associated protein 2-4
- Gene Aliases
- KAP2.1B, KAP2.4, KRTAP2.4
- Chromosome Location
- Chr. 17: 41065116 - 41065879 on Build GRCh38
- UniGene ID
- Hs.560502
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ406929.1 |
- |
|
389 |
|
View Full Product Details