Gene Transcripts

Gene Symbol
NOTCH2NL
Entrez Gene ID
388677
Gene Name
notch 2 N-terminal like
Gene Aliases
N2N
Chromosome Location
Chr. 1: 146151908 - 146229032 on Build GRCh38
UniGene ID
Hs.655156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203458.4 NP_982283.2 5 1368
GenBank mRNA BC065219.1 - 5 1320
BX537434.1 - 6 1447

Gene Symbol
LOC100996717
Entrez Gene ID
100996717
Gene Name
notch homolog 2 N-terminal-like protein
Gene Aliases
Chromosome Location
Chr. 1: 149386823 - 149475116 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003051.1 XP_016858540.1 5 1329

Gene Symbol
LOC100996763
Entrez Gene ID
100996763
Gene Name
notch homolog 2 N-terminal-like protein
Gene Aliases
Chromosome Location
Chr. 1: 148596477 - 148712497 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006711700.3 XP_006711763.1 5 1277
XM_006711703.3 XP_006711766.1 5 1339

View Full Product Details