Gene Transcripts

Gene Symbol
OR10H5
Entrez Gene ID
284433
Gene Name
olfactory receptor family 10 subfamily H member 5
Gene Aliases
OR19-25, OR19-26
Chromosome Location
Chr. 19: 15794049 - 15794996 on Build GRCh38
UniGene ID
Hs.553676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004466.1 NP_001004466.1 1 524
GenBank mRNA BC136869.1 - 1 622

View Full Product Details