Gene Transcripts
- Gene Symbol
- CDRT15
- Entrez Gene ID
- 146822
- Gene Name
- CMT1A duplicated region transcript 15
- Gene Aliases
- Chromosome Location
- Chr. 17: 14235856 - 14236833 on Build GRCh38
- UniGene ID
- Hs.735172
- Species
- Homo sapiens
- Gene Symbol
- CDRT15P1
- Entrez Gene ID
- 94158
- Gene Name
- CMT1A duplicated region transcript 15 pseudogene 1
- Gene Aliases
- CDRT15P
- Chromosome Location
- Chr. 17: 14024498 - 14025598 on Build GRCh38
- UniGene ID
- Hs.633073
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003261.1 |
|
1 |
252 |
|
GenBank mRNA |
AA453375.1 |
- |
1 |
253 |
|
BX118788.1 |
- |
1 |
253 |
|
View Full Product Details