Gene Transcripts

Gene Symbol
PRAMEF4
Entrez Gene ID
400735
Gene Name
PRAME family member 4
Gene Aliases
Chromosome Location
Chr. 1: 12879212 - 12886201 on Build GRCh38
UniGene ID
Hs.551617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009611.4 NP_001009611.2 2 343
GenBank mRNA AL049681.1 - 2 343

View Full Product Details