Gene Transcripts

Gene Symbol
NBPF8
Entrez Gene ID
728841
Gene Name
neuroblastoma breakpoint family member 8
Gene Aliases
NBPF8P
Chromosome Location
Chr. 1: 120436351 - 120467844 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY894567.1 - 1 293
AY894568.1 - 2 191
AY894571.1 - 1 293
AY894572.1 - 1 293
AY894573.1 - 1 293
BC086308.1 - 1 136 5453529

View Full Product Details