Gene Transcripts

Gene Symbol
Lrrc42
Entrez Gene ID
298309
Gene Name
leucine rich repeat containing 42
Gene Aliases
RGD1308919
Chromosome Location
Chr. 5: 126835170 - 126856283 on Build Rnor_6.0
UniGene ID
Rn.98323
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025653.1 NP_001020824.1 8 1330
XM_006238503.2 XP_006238565.1 8 1252
GenBank mRNA BC098685.1 - 8 1330

View Full Product Details