Gene Transcripts

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
Chromosome Location
Chr. 15: 75674322 - 75712848 on Build GRCh38
UniGene ID
Hs.513044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001897.4 NP_001888.2 3 959
GenBank mRNA AY359468.1 - 3 866
X96753.1 - 3 866

View Full Product Details