Gene Transcripts

Gene Symbol
Tmc4
Entrez Gene ID
308310
Gene Name
transmembrane channel-like 4
Gene Aliases
Chromosome Location
Chr. 1: 64114687 - 64126576 on Build Rnor_6.0
UniGene ID
Rn.138375
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034104.1 NP_001029276.1 616
GenBank mRNA BC100657.1 - 616

Gene Symbol
LOC103690093
Entrez Gene ID
103690093
Gene Name
transmembrane channel-like protein 4
Gene Aliases
Chromosome Location
Chr. 1: 64046203 - 64054860 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758838.1 XP_008757060.1 4 800

View Full Product Details