Gene Transcripts

Gene Symbol
Stra6
Entrez Gene ID
363071
Gene Name
stimulated by retinoic acid 6
Gene Aliases
RGD1307551
Chromosome Location
Chr. 8: 62925364 - 62944438 on Build Rnor_6.0
UniGene ID
Rn.12735
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029924.1 NP_001025095.1 4 404
GenBank mRNA BC097373.1 - 4 404

View Full Product Details