Gene Transcripts

Gene Symbol
Coq9
Entrez Gene ID
498909
Gene Name
coenzyme Q9
Gene Aliases
Chromosome Location
Chr. 19: 10583855 - 10596848 on Build Rnor_6.0
UniGene ID
Rn.22044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035257.1 NP_001030334.1 7 824
GenBank mRNA BC079370.1 - 7 810
BC104702.1 - 7 810
BC127449.1 - 7 825 8365327

View Full Product Details