Gene Transcripts

Gene Symbol
Olfr44
Entrez Gene ID
18343
Gene Name
olfactory receptor 44
Gene Aliases
IB3, MOR171-5
Chromosome Location
Chr. 9: 39484087 - 39493988 on Build GRCm38
UniGene ID
Mm.246538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146830.2 NP_667041.2 2 479
GenBank mRNA AF102518.1 - 1 258
CB174038.1 - 2 486

View Full Product Details