Gene Transcripts

Gene Symbol
Taar6
Entrez Gene ID
215855
Gene Name
trace amine-associated receptor 6
Gene Aliases
Gm228, taR-6
Chromosome Location
Chr. 10: 23984609 - 23985646 on Build GRCm38
UniGene ID
Mm.377237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010828.1 NP_001010828.1 1 103

View Full Product Details