Gene Transcripts

Gene Symbol
Brsk2
Entrez Gene ID
75770
Gene Name
BR serine/threonine kinase 2
Gene Aliases
4833424K13Rik, SAD-A, SADA
Chromosome Location
Chr. 7: 141949751 - 142004244 on Build GRCm38
UniGene ID
Mm.274868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009929.3 NP_001009929.1 4, 5 747
NM_001009930.3 NP_001009930.1 4, 5 747
NM_001276763.1 NP_001263692.1 4, 5 747
NM_029426.2 NP_083702.1 4, 5 747
GenBank mRNA AK158574.1 - 4, 5 496
AK173268.1 - 3, 4 322
AY533672.1 - 4, 5 412
AY533673.1 - 4, 5 412
AY533674.1 - 4, 5 412
AY660739.1 - 4, 5 747
BC056498.1 - 1, 2 23
JF707771.1 - 4, 5 412
JF707772.1 - 4, 5 412

View Full Product Details