Gene Transcripts

Gene Symbol
Snx25
Entrez Gene ID
102141
Gene Name
sorting nexin 25
Gene Aliases
AI661919, Gm1699, Gm173, Sbbi31
Chromosome Location
Chr. 8: 46033261 - 46151858 on Build GRCm38
UniGene ID
Mm.267258
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207213.2 NP_997096.2 11 1669
GenBank mRNA AY601646.1 - 10 1888
BC054725.1 - 6 925 5699137
BC057063.1 - 10 1417

View Full Product Details