Gene Transcripts

Gene Symbol
RGMA
Entrez Gene ID
56963
Gene Name
repulsive guidance molecule family member a
Gene Aliases
RGM
Chromosome Location
Chr. 15: 93043406 - 93089214 on Build GRCh38
UniGene ID
Hs.271277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166283.1 NP_001159755.1 4 2610
NM_001166286.1 NP_001159758.1 3 2470
NM_001166287.1 NP_001159759.1 4 2584
NM_001166288.1 NP_001159760.1 4 2468
NM_001166289.1 NP_001159761.1 4 2488
NM_020211.2 NP_064596.2 4 2637
GenBank mRNA AK074910.1 - 4 2627
AK074966.1 - 3 2444
AK074980.1 - 4 2404
AK125047.1 - 1 2997
AK125204.1 - 5 2593
AL136826.1 - 4 2451
AL390083.1 - 3 2393
BC015886.1 - 3 2461

View Full Product Details