Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320395.1 | NP_001307324.1 | 4 | 1701 | |
NM_001320397.1 | NP_001307326.1 | 5 | 1658 | ||
NM_001320398.1 | NP_001307327.1 | 5 | 1754 | ||
NM_001320399.1 | NP_001307328.1 | 6 | 1883 | ||
NM_001320400.1 | NP_001307329.1 | 3 | 1626 | ||
NM_001320401.1 | NP_001307330.1 | 4 | 1682 | ||
NM_020357.2 | NP_065090.1 | 5 | 1757 | ||
NR_135223.1 | 5 | 1705 | |||
NR_135224.1 | 4 | 1542 | |||
NR_135225.1 | 5 | 1702 | |||
NR_135226.1 | 3 | 1574 | |||
NR_135227.1 | 5 | 1958 | |||
NR_135228.1 | 4 | 1649 | |||
NR_135229.1 | 5 | 1886 | |||
NR_135230.1 | 4 | 1630 | |||
NR_135231.1 | 3 | 1467 | |||
NR_135232.1 | 6 | 1800 | |||
XM_017006922.1 | XP_016862411.1 | 4 | 1612 | ||
GenBank mRNA | AB037675.1 | - | 5 | 1653 | |
AK025551.1 | - | 5 | 1597 | ||
AK098760.1 | - | 5 | 1636 | ||
AL110143.1 | - | 1 | 227 | ||
BC022001.2 | - | 5 | 1655 | 4823990 | |
BC107796.1 | - | 5 | 1599 | ||
BX647886.1 | - | 1 | 2344 | ||
CR627371.1 | - | 4 | 1442 |