Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020485.4 | NP_065231.3 | 10 | 1463 | |
NM_138616.3 | NP_619522.3 | 7 | 1010 | ||
NM_138617.3 | NP_619523.3 | 7 | 1068 | ||
NM_138618.3 | NP_619524.3 | 9 | 1329 | ||
XM_005245957.3 | XP_005246014.1 | 8 | 1774 | ||
XM_011541888.2 | XP_011540190.1 | 11 | 1963 | ||
XM_011541889.2 | XP_011540191.1 | 10 | 1478 | ||
XM_011541890.2 | XP_011540192.1 | 9 | 1844 | ||
XM_011541891.2 | XP_011540193.1 | 11 | 1580 | ||
XM_017002014.1 | XP_016857503.1 | 8 | 1723 | ||
GenBank mRNA | AF056965.1 | - | 10 | 1375 | |
AF510065.1 | - | 10 | 1419 | ||
AF510066.1 | - | 10 | 1419 | ||
AF510067.1 | - | 10 | 1419 | ||
AF510068.1 | - | 10 | 1419 | ||
BC139905.1 | - | 10 | 1505 | 40147407 | |
JX420291.1 | - | 10 | 1377 | ||
JX420292.1 | - | 10 | 1377 | ||
JX420293.1 | - | 10 | 1377 | ||
JX420294.1 | - | 10 | 1377 | ||
JX420295.1 | - | 10 | 1377 | ||
JX420296.1 | - | 10 | 1377 | ||
JX420297.1 | - | 10 | 1377 | ||
JX420298.1 | - | 10 | 1377 | ||
JX420299.1 | - | 10 | 1377 | ||
JX420300.1 | - | 10 | 1377 | ||
JX420301.1 | - | 10 | 1377 | ||
JX420302.1 | - | 10 | 1377 | ||
X54534.1 | - | 10 | 1426 | ||
X63095.1 | - | 7 | 990 | ||
X63096.1 | - | 7 | 930 |