Gene Transcripts

Gene Symbol
OR2A14
Entrez Gene ID
135941
Gene Name
olfactory receptor family 2 subfamily A member 14
Gene Aliases
OR2A14P, OR2A6, OST182
Chromosome Location
Chr. 7: 144129113 - 144130045 on Build GRCh38
UniGene ID
Hs.534547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001659.1 NP_001001659.1 1 253
GenBank mRNA BC136889.1 - 1 308

View Full Product Details