Gene Transcripts

Gene Symbol
KRTAP12-4
Entrez Gene ID
386684
Gene Name
keratin associated protein 12-4
Gene Aliases
KRTAP12.4
Chromosome Location
Chr. 21: 44654213 - 44654659 on Build GRCh38
UniGene ID
Hs.689664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198698.1 NP_941971.1 1 277
GenBank mRNA AB076360.1 - 1 277
BC125198.1 - 1 277 40123223

View Full Product Details