Gene Transcripts

Gene Symbol
Ang
Entrez Gene ID
305843
Gene Name
angiogenin, ribonuclease, RNase A family, 5
Gene Aliases
Ang1
Chromosome Location
Chr. 15: 28022908 - 28028636 on Build Rnor_6.0
UniGene ID
Rn.217453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006992.1 NP_001006993.1 1 161
XM_006251874.2 XP_006251936.1 2 344
GenBank mRNA AY395869.1 - 1 161

View Full Product Details