Gene Transcripts

Gene Symbol
Ly49i4
Entrez Gene ID
494204
Gene Name
Ly49 inhibitory receptor 4
Gene Aliases
Chromosome Location
Chr. 4: 164515105 - 164536581 on Build Rnor_6.0
UniGene ID
Rn.83631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009495.1 NP_001009495.1 1 166
XM_008763405.1 XP_008761627.1 2 281
GenBank mRNA AY649834.1 - 1 166

View Full Product Details