Gene Transcripts

Gene Symbol
Defb11
Entrez Gene ID
641630
Gene Name
defensin beta 11
Gene Aliases
Chromosome Location
Chr. 16: 75146220 - 75156555 on Build Rnor_6.0
UniGene ID
Rn.155421
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037505.1 NP_001032594.1 2 186
GenBank mRNA AY621343.1 - 2 186

View Full Product Details