Gene Transcripts

Gene Symbol
Hist3h2a
Entrez Gene ID
64646
Gene Name
histone cluster 3, H2a
Gene Aliases
H2a
Chromosome Location
Chr. 10: 45297603 - 45299155 on Build Rnor_6.0
UniGene ID
Rn.198869
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021840.2 NP_068612.2 1 709
GenBank mRNA BC099140.1 - 1 709

View Full Product Details