Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_212510.1 | NP_997675.1 | 604 | ||
GenBank mRNA | BC058498.1 | - | 604 | ||
FQ209874.1 | - | 445 | |||
FQ209881.1 | - | 598 | |||
FQ209956.1 | - | 604 | |||
FQ216877.1 | - | 597 | |||
FQ221165.1 | - | 599 | |||
FQ221243.1 | - | 598 | |||
FQ221413.1 | - | 598 | |||
FQ221458.1 | - | 598 | |||
FQ221831.1 | - | 605 | |||
FQ222162.1 | - | 598 | |||
FQ222289.1 | - | 598 | |||
FQ222457.1 | - | 598 | |||
FQ222474.1 | - | 602 | |||
FQ222662.1 | - | 598 | |||
FQ223818.1 | - | 599 | |||
FQ228440.1 | - | 598 |