Gene Transcripts
- Gene Symbol
- SV2C
- Entrez Gene ID
- 22987
- Gene Name
- synaptic vesicle glycoprotein 2C
- Gene Aliases
- Chromosome Location
- Chr. 5: 75847463 - 76353939 on Build GRCh38
- UniGene ID
- Hs.646953
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297716.1 |
NP_001284645.1
|
7, 8 |
1443 |
|
NM_014979.3 |
NP_055794.3
|
7, 8 |
1443 |
|
XM_011543281.2 |
XP_011541583.1
|
7, 8 |
1810 |
|
XM_011543282.2 |
XP_011541584.2
|
7, 8 |
2370 |
|
XM_017009244.1 |
XP_016864733.1
|
3, 4 |
361 |
|
GenBank mRNA |
AB028977.1 |
- |
5, 6 |
504 |
|
AK094917.1 |
- |
7, 8 |
1444 |
|
BC100824.1 |
- |
7, 8 |
1379 |
40001296 |
BC100825.1 |
- |
7, 8 |
1377 |
40001297 |
BC100826.1 |
- |
7, 8 |
1377 |
40001302 |
BC100827.1 |
- |
7, 8 |
1377 |
40001303 |
View Full Product Details