Gene Transcripts

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
Chromosome Location
Chr. 5: 95555108 - 95605102 on Build GRCh38
UniGene ID
Hs.585051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198150.2 NP_937793.1 5 902
XM_005271904.3 XP_005271961.1 6 966
GenBank mRNA AK303855.1 - 5 954
AK316253.1 - 5 921
AL832711.1 - 4 585
AY358596.1 - 5 862
AY875939.1 - 5 744
BC036047.1 - 5 764
BC050392.1 - 6 912
BC130329.1 - 5 840 40146360
BC144161.1 - 5 840

View Full Product Details