Gene Transcripts

Gene Symbol
NRK
Entrez Gene ID
203447
Gene Name
Nik related kinase
Gene Aliases
NESK
Chromosome Location
Chr. X: 105821944 - 105958610 on Build GRCh38
UniGene ID
Hs.209527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198465.3 NP_940867.2 13 2039
XM_006724632.3 XP_006724695.1 13 2001
XM_006724633.3 XP_006724696.1 13 2001
XM_006724634.3 XP_006724697.1 13 2001
XM_011530887.2 XP_011529189.1 13 1793
GenBank mRNA AK131549.1 - 13 3065
BX538345.1 - 13 2041

View Full Product Details