Gene Transcripts

Gene Symbol
ENOPH1
Entrez Gene ID
58478
Gene Name
enolase-phosphatase 1
Gene Aliases
E1, MASA, MST145, mtnC
Chromosome Location
Chr. 4: 82430480 - 82461175 on Build GRCh38
UniGene ID
Hs.18442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292017.1 NP_001278946.1 2, 3 508
NM_021204.4 NP_067027.1 2, 3 537
GenBank mRNA AF087881.1 - 2, 3 407
AF113125.1 - 2, 3 408
AK022656.1 - 2, 3 403
BC065815.1 - 2, 3 375 6526329
CN335976.1 - 2, 3 349
CR457141.1 - 2, 3 187
DA067292.1 - 2, 3 537
DA347866.1 - 2, 3 444

View Full Product Details